Skip to Content
Merck
All Photos(2)

Documents

MABE901

Sigma-Aldrich

Anti-TAF15 Antibody, clone 4D71

clone 4D71, from mouse

Synonym(s):

TATA-binding protein-associated factor 2N, 68 kDa TATA-binding protein-associated factor, TAF(II)68, TAFII68, RNA-binding protein 56

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D71, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TAF15(8148)

General description

TAF15, also known as TATA-binding protein-associated factor 2N, or 68 kDa TATA-binding protein-associated factor, or TAF(II)68, TAFII68, or RNA-binding protein 56, and encoded by the gene TAF15/RBP56/TAF2N, is a RNA and ssDNA binding protein important in transcription initiation. TAF15 aligns with the RNA polymerase II (Pol II) multiprotein complex that together with other proteins like TBP initiates transcription. TAF15 is ubiquitously expressed, and TAF15 shuttles from the nucleus to the cytoplasm upon activation; methylation of TAF15 appears to guide nuclear localization and transcriptional activity. Translocations involving the TAF15 gene may play a role in acute leukemia and extraskeletal myxoid chondrosarcoma, and mutations in this gene are associated with amyotrophic lateral sclerosis (ALS) diseases.

Immunogen

Recombinant protein corresponding to human TAF15.

Application

Anti-TAF15 Antibody, clone 4D71 is an antibody against TAF15 for use in western blotting & ICC.
Immunocytochemistry Analysis: A 1:500 to a 1:1,000 dilution from a representative lot from an independent laboratory detected TAF15 in HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 μg/mL of this antibody detected TAF15 in 10 μg of HeLa cell lysate.

Target description

~68 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service