Skip to Content
Merck
All Photos(2)

Documents

MABE340-AF488

Sigma-Aldrich

Anti-MDM2 Antibody, clone IF2, Alexa Fluor 488 Conjugate

clone IF2, from mouse, ALEXA FLUOR 488

Synonym(s):

E3 ubiquitin-protein ligase Mdm2, Double minute 2 protein, Hdm2, Oncoprotein Mdm2, p53-binding protein Mdm2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IF2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)

General description

E3 ubiquitin-protein ligase Mdm2 (UniProt Q00987; also known as Double minute 2 protein, Hdm2, Oncoprotein Mdm2, p53-binding protein Mdm2) is encoded by the MDM2 gene (Gene ID 4193) in human. MDM2 is an E3 ubiquitin ligase that is most widely studied for its role in regulating the p53 tumor suppressor. MDM2 antagonizes p53 transcriptional activity by binding to and blocking p53 N-terminal trans-activation domain. MDM2 can also monoubiquitinate p53, thereby promoting p53 cytoplasmic localization to prevent p53 nuclear funtion. In addition, MDM2, when in complex with p300, also mediates p53 polyubiquitination, resulting in p53 proteosomal degradation. MDM2 overexpression is shown to inhibit DNA double-strand break repair mediated through its interaction with Nbs1 in a p53-independent manner.

Specificity

Clone IF2 recognizes the ~85/90 kDa (apparent MW) MDM2, as well as isoforms of ~57/60 kDa and ~74/76 kDa by immunoblotting using the unconjugated antibody (Cat. Nos. MABE340 & OP46).

Immunogen

Epitope: Within amino acids 26-169.
Recombinant human MDM2.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MDM2 in A431 cells.
The unconjugated antibody (Cat. Nos. MABE340 & OP46) is shown to be suitable also for immunohistochemistry and Western blotting applications.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Apoptosis - Additional
This Anti-MDM2 Antibody, clone IF2, Alexa Fluor 488 Conjugate is validated for use in Immunocytochemistry for the detection of MDM2.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected MDM2 in HeLa cells.

Target description

~85 kDa observed.

Physical form

Protein G purified.
Purified mouse monoclonal IgG2b antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service