Skip to Content
Merck
All Photos(1)

Key Documents

MABE254

Sigma-Aldrich

Anti-Protein-arginine deiminase 4 Antibody, clone 10F1

clone 10F1, from mouse

Synonym(s):

HL-60 PAD, Protein-arginine deiminase type-4, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10F1, monoclonal

species reactivity

human

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PADI4(23569)

General description

Protein-arginine deiminase type-4 (PAD4; formerly PAD5) is one of five PAD (1-4, 6) homologues that belong to the superfamily of multidomain amidinotransferases. The PAD enzymes catalyze deiminating reactions in which guanidinium is hydrolyzed to peptidylcitrulline. PAD4 is a calcium-regulated nuclear-localized enzyme. It modifies arginine residues on histones H2A, H3, and H4 (H2 Arg3; H3 Arg2, H3 Arg8, H3 Arg17, H3 Arg26; and H4 Arg3). Several studies have suggested that PAD4-mediated citrullination has important implications for gene regulation. PAD4 may perform deimination at sites overlapping with arginine methylation, and antagonize the transcription-promoting effects of methylating enzymes such as CARM1 and PRMT1. PAD4 may therefore inhibit gene expression, most notably expression of cell-cycle regulating p53 genes such as p21. Previous studies have suggested that PAD4 may play central role in the development of cancer.

Immunogen

GST-tagged recombinant protein corresponding to human Protein-arginine deminiase 4.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Use Anti-Protein-arginine deiminase 4 Antibody, clone 10F1 (mouse monoclonal antibody) validated in WB, IP to detect PAD-4 also known as HL-60 PAD, Protein-arginine deiminase type-4, Peptidylarginine deiminase IV.

Quality

Evaluated by Western Blot in DMSO treated and untreated HL-60 cell lysates.

Western Blot Analysis: 0.25 µg/mL of this antibody detected Protein-arginine deminiase 4 in 10 µg DMSO treated and untreated HL-60 cell lysates.

Target description

~74 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
DMSO treated and untreated HL-60 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yebin Zhou et al.
Frontiers in immunology, 8, 1200-1200 (2017-10-11)
Autoantibodies directed against citrullinated epitopes of proteins are highly diagnostic of rheumatoid arthritis (RA), and elevated levels of protein citrullination can be found in the joints of patients with RA. Calcium-dependent peptidyl-arginine deiminases (PAD) are the enzymes responsible for citrullination.
Yebin Zhou et al.
Scientific reports, 8(1), 15228-15228 (2018-10-17)
Neutrophils are critical for the defense against pathogens, in part through the extrusion of extracellular DNA traps, phagocytosis, and the production of reactive oxygen species. Neutrophils may also play an important role in the pathogenesis of rheumatoid arthritis (RA) through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service