Skip to Content
Merck

Skip To

MABE1007

Anti-TEL-AML1 fusion Antibody, clone 6F2

Synonym(s):

TEL-AML1 fusion|ETV6-RUNX1|Transcription factor ETV6|ETS translocation variant 6|ETS-related protein Tel1|Tel|Runt-related transcription factor 1|Acute myeloid leukemia 1 protein|Core-binding factor subunit alpha-2|CBF-alpha-2|Oncogene AML-1|Polyomavirus

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
monoclonal
Species reactivity:
human
Application:
ChIP, FACS, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

clone

monoclonal

species reactivity

human

technique(s)

ChIP: suitable, flow cytometry: suitable, western blot: suitable

General description

The chimeric transcription factor TEL-AML1 (ETV6-RUNX1) is the result of the reciprocal translocation t(12;21)(p13;q22), the most common structural genomic alteration found in childhood B-cell precursor acute lymphoblastic leukaemia (ALL). The runt-homology DNA-binding domain of AML1 retained in the TEL-AML1 mediates the DNA-binding activity of the fusion protein. The fusion protein not only exhibits altered transcription activity, it also affects normal transcription activity by directly competing against wild-type AML1 for promoter binding, sequestering transcriptional cofactors available for the wild-type protein, as well as dimerizing with wild-type protein.

Immunogen

KLH-conjugated linear peptide corresponding to human TEL-AML1 fusion.

Application

Anti-TEL-AML1 fusion Antibody, clone 6F2 is an antibody against TEL-AML1 fusion for use in Western Blotting, Flow Cytometry, Chromatin Immunoprecipitation .
Flow Cytometry Analysis: A representative lot detected TEL-AML1 fusion induction upon mifepristone treatment in BA/F3 cells expressing inducible TEL-AML1 fusion (Linka, Y., et al. (2013). Blood Cancer J. 3:e151).
Chromatin Immunoprecipitation Analysis

Analysis Note

Evaluated by Western Blotting in lysate from mifepristone-treated murine BA/F3 cells expressing inducible TEL-AML1 fusion.
Western Blotting Analysis: A 1:20,000 dilution of this antibody detected TEL-AML1 fusion induction in 2.5 ug lysate from mifepristone-treated murine BA/F3 cells expressing inducible TEL-AML1 fusion.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

A major focus of breast cancer research is to understand the mechanisms responsible for disease progression and drug resistance. Toward that end, it has been found that approximately two thirds of all human breast carcinomas overexpress the Estrogen Receptor α (ERα) protein and it remains the primary pharmacological target for endocrine therapy1,2. The normal cellular function of ERα is as a transcription factor that mediates a wide variety of physiological processes, many of which are dependent upon phosphorylation of the receptor at specific amino acid residues3,4. Indeed, ERα is known to be phosphorylated at a multitude of different sites, yet how these all correlate to disease remains unclear5. Here, we interrogated multiple sites of ERα for phosphorylation status by screening an extensive panel of different breast cancer patient samples and other non-breast cancer tissue microarray (TMA) slide samples to determine their relevance to disease.

Historically chromatin was considered to contain only DNA, histones, non-histone proteins and transiently associated mRNAs. While this remains true, current data suggest that a variety of non-coding RNAs (e.g. long non-coding RNAs, enhancer RNAs and even miRNAs) also associate with chromatin and serve regulatory functions.






Questions

Reviews

No rating value

Active Filters