Skip to Content
Merck
All Photos(2)

Key Documents

MABC949

Sigma-Aldrich

Anti-Glycoprotein 78 Antibody, clone 3F3A

clone 3F3A, from rat

Synonym(s):

E3 ubiquitin-protein ligase AMFR, AMF receptor, Autocrine motility factor receptor, gp78, RING finger protein 45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
3F3A, monoclonal
application:
ICC
IHC
WB
species reactivity:
mouse, human
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
citations:
1
Pricing and availability is not currently available.

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F3A, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GPR78(27201)

General description

E3 ubiquitin-protein ligase AMFR (UniProt Q9UKV5; also known as AMF receptor, Autocrine motility factor receptor, gp78, RING finger protein 45) is encoded by the RNF45 (also known as AMFR) gene (Gene ID 267) in human. Glycoprotein 78 (gp78) is an endoplasmic reticulum (ER) membrane-anchored E3 ubiquitin ligase that plays a key role in ER-associated degradation (ERAD). Gp78 is localized to a mitochondria-associated ER domain, where it induces mitochondrial fragmentation by targeting the mitochondrial fusion proteins mitofusin 1 and 2 (Mfn1 and Mfn2, respectively) for degradation upon mitochondrial depolarization. Other known ERAD substrates processed by gp78 include ApoB lipoprotein, HMG CoA reductase, CD3-delta, cystic fibrosis transmembrane conductance regulator, the metastasis suppressor KAI1, unglycosylated prion protein (PrP), and the T-cell receptor. Gp78 is a 7-transmembrane protein (a.a. 82-102, 122-142, 145-165, 186-206, 215-235, 276-296, 429-449) with a RING-type Zinc finger (a.a. 341-379), a CUE domain (a.a. 456-498), and a VCP/p97-interacting motif (VIM; a.a. 622-640).

Specificity

Clone 3F3A detected glycoprotein 78 (gp78) and a ~150 kDa glycoprotein in B16-F1 mouse melanoma cell extract. Cell extract sialidase treatment decreased the antibody′s immunoreactivity toward gp78 and abolished ~150 kDa band detection (Nabi, I.R., et al. (1990). Cancer Res. 50(2):409-414).

Immunogen

Epitope: extracellular domain
Peanut lectin (PNA) resin-purified membrane glycoprotein 78 from B16-F1 cells (Nabi, I.R., and Raz, A. (1987). Int. J. Cancer. 40(3):396-402).

Application

Detect GP78 using this rat monoclonal Anti-Glycoprotein 78, clone 3F3A, Cat. No. MABC949, validated for use in Function Assays, Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Immunocytochemistry: A representative lot detected glycoprotein 78 immunoreactivity colocalized with that of the mitochondria marker OxPhosV in COS-7 cells (Courtesy of I. Robert Nabi, Ph.D., University of British Columbia, Canada).

Affects Function: A representative lot stimulated the mobility of B16-F1 mouse melanoma cells on colloidal gold-coated surface (Nabi, I.R., et al. (1990). Cancer Res. 50(2):409-414).

Immunocytochemistry Analysis: A representative lot detected both surface and cytoplasmic gp78 immunoreactivity by fluorescent immunocytochemistry staining of intact or MeOH/3% paraformaldehyde-fixed and 0.5% Triton X-100-permeablized mouse A31 fibroblasts and A31M angiosarcoma cells (Niinaka, Y., et al. (1998). Cancer Res. 58(12):2667-2674).

Immunocytochemistry Analysis: Clone 3F3A ascites fluid immunolocalized gp78 on the surface of adherent B16-F1 mouse melanoma cells by fluorescent immunocytochemistry staining of 3% paraformaldehyde-fixed cells (Nabi, I.R., et al. (1990). Cancer Res. 50(2):409-414).

Immunohistochemistry Analysis: A representative lot detected an elevated gp78 immunoreactivity in frozen mammary gland sections from 235RLF transgenic mice than age-matched wild type mice (Joshi, B., et al. (2010). J. Biol. Chem. 285(12):8830-8839).

Western Blotting Analysis: A representative lot detected shRNA-mediated gp78 downregulation in HT-1180 human fibrosarcoma cells (Fu, M., et al. (2013). Mol. Biol. Cell. 24(8):1153-1162).

Western Blotting Analysis: A representative lot detected endogenous gp78 and gp78 transgene expression in mammary glands from wild-type and transgenic mice. Clone 3F3A detected a higher gp78 expression in metastatic MDA-435 cells than in non-metastatic MCF7 breast carcinoma cells (Joshi, B., et al. (2010). J. Biol. Chem. 285(12):8830-8839).

Western Blotting Analysis: A representative lot detected much higher gp78 expression in angiosarcoma (human HT-1180 and mouse A31M) than in fibroblast (human IMR90 and mouse A31) lines (Niinaka, Y., et al. (1998). Cancer Res. 58(12):2667-2674).

Western Blotting Analysis: A representative lot detected gp78 and a ~150 kDa band in B16-F1 mouse melanoma cell extract. Autocrine motility factor (AMF) competed against clone 3F3A for binding the gp78 target band. Cell extract sialidase treatment decreased the antibody′s immunoreactivity toward gp78 and abolished ~150 kDa band detection (Nabi, I.R., et al. (1990). Cancer Res. 50(2):409-414).
Research Category
Apoptosis & Cancer

Quality

Western Blotting Analysis: A 1:62.5 dilution of this antibody detected glycoprotein 78 (gp78) in 50 µg of HeLa cell lysate.

Target description

~78 kDa observed. Target band size appears larger than the calculated molecular weight of 73.00 kDa due to glycosylation. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified rat IgM in buffer containing PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ameair Abu Irqeba et al.
PloS one, 15(12), e0243075-e0243075 (2020-12-02)
Prenylated Rab Acceptor 1 (PRA1/Rabac1) is a four-pass transmembrane protein that has been found to localize to the Golgi and promiscuously associate with a diverse array of Rab GTPases. We have previously identified PRA1 to be among the earliest significantly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service