Skip to Content
Merck
All Photos(1)

Key Documents

MABC779

Sigma-Aldrich

Anti-Folate, clone 8/33 Antibody

clone 8/33, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8/33, monoclonal

species reactivity (predicted by homology)

all

technique(s)

ELISA: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

Immunogen

KLH-conjugated Tetrahydrofolic acid corresponding to Folate.

Application

Anti-Folate, clone 8/33 | MABC779 is an antibody against Folate for use in ELISA.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by ELISA in Tetrahydrofolic acid-BSA.

ELISA Analysis: 0.1-100 µg/mL of this antibody detected Folate in Tetrahydrofolic acid-BSA.

Linkage

Replaces: CBL65

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service