MABC42
Anti-Pleiotrophin Antibody, clone 3B10
clone 3B10, from mouse
Synonym(s):
pleiotrophin, neurite growth-promoting factor 1, Heparin-binding brain mitogen, heparin binding growth factor 8, Heparin-binding growth-associated molecule, heparin affin regulatory protein, Heparin-binding neurite outgrowth-promoting factor 1, pleiotrop
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
3B10, monoclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PTN(5764)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Developmental Neuroscience
Quality
Western Blot Analysis: 0.5 µg/mL of this antibody detected Pleiotrophin in 10 µg of human placenta tissue lysate.
Target description
Linkage
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Human placenta tissue lysate
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service