Skip to Content
Merck
All Photos(1)

Key Documents

MABC1736

Sigma-Aldrich

Anti-CIDE-A Antibody, clone V62P1E3 B10

Synonym(s):

Cell death activator CIDE-A;Cell death-inducing DFFA-like effector A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

V62P1E3 B10, monoclonal

mol wt

calculated mol wt 24.69 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CIDEA(1149)

General description

Cell death activator CIDE-A (UniProt: O60543; also known as Cell death-inducing DFFA-like effector A) is encoded by the CIDEA gene (Gene ID: 1149) in human. Three members of the CIDE family of proteins have been described in human CIDE-A, CIDE-B, and CIDE-C. They all contain a common N-terminal CIDE-N domain and a C-terminal CIDE-C domain. CIDE-A is a brown fat-associated protein that has been implicated in lipid and energy metabolism. Its expression has been reported in colon and normal breast tissue. It binds to lipid droplets and regulates their enlargement, thereby restricting lipolysis and favoring storage. It is shown to be enriched at lipid droplet contact sites and at focal contact sites between lipid droplets where it promotes directional net neutral lipid transfer from the smaller to larger lipid droplets. It is reported to strongly promote triglyceride accumulation even in cell types that do not normally store large amounts of neutral lipid. Its expression levels in adipose tissue have been correlated with insulin sensitivity, which could be related to enhanced storage of triglyceride resulting in reduced levels of fatty acid levels in the circulation. CIDE-A is also reported to acts as a CCAAT Enhancer Binding Protein b (CEBPB) coactivator in mammary epithelial cells and controls the expression of CEBPB downstream target genes, such as ID2, IGF1, PRLR, SOCS1, SOCS3, XDH. Higher expression of CIDE-A has been reported in colorectal tumors compared to normal colonic mucosa. (Ref.: Alnabulsi, A., et al. (2019). Int. J. Cancer. 145(4);1138-1147; Puri, V., et al. (2008). Proc. Natl. Acad. Sci. USA. 105(22); 7833-7838).

Specificity

Clone V62P1E3 B10 is a mouse monoclonal antibody that detects human Cell death activator CIDE-A. It targets an epitope within 9 amino acids from the C-terminal region.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 9 amino acids from the C-terminal region of human CIDE-A.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human colon cancer tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected CIDE-A in human colon cancer tissue sections.

Tested Applications


Western Blotting Analysis: A representative lot detected CIDE-A in Western Blotting applications (Alnabulsi, A., et al. (2019). Int J Cancer. 145(4):1138-1147).

ELISA Analysis: A representative lot detected CIDEA in ELISA applications (Alnabulsi, A., et al. (2019). Int J Cancer. 145(4):1138-1147).

Immunohistochemistry Applications: A representative lot detected CIDEA in Immunohistochemistry applications (Alnabulsi, A., et al. (2019). Int J Cancer. 145(4):1138-1147).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CIDE-A, clone V62P1E3 B10, Cat. No. MABC1736, is a mouse monoclonal antibody that detects CIDE-A and is tested for use in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service