Skip to Content
Merck
All Photos(3)

Key Documents

MABC1608

Sigma-Aldrich

Anti-Mucin-16 (CA-125) Antibody, clone 5E11

clone 5E11, from mouse

Synonym(s):

MUC-16, Ovarian cancer-related tumor marker CA125, CA-125, Ovarian carcinoma antigen CA125

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E11, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
proximity ligation assay: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MUC16(94025)

General description

Mucin-16 (UniProt: Q8WXI7; also known as MUC-16, Ovarian cancer-related tumor marker CA125, CA-125, Ovarian carcinoma antigen CA125) is encoded by the MUC16 (also known as CA125) gene (Gene ID: 94025) in human. Mucin-16 is a heavily O-glycosylated protein that can be liberated into the extracellular space following the phosphorylation of the intracellular C-terminus, which induces proteolytic cleavage and liberation of extracellular domain. I provides protective, lubricating barrier against particles and infectious agents at mucosal surfaces. Mucin-16 is composed of three domains: a Serine, Threonine-rich N-terminal domain, a repeated domain containing between 12 and 60 partially conserved tandem repeats of 156 amino acids and a C-terminal transmembrane domain with a short cytoplasmic tail. Its expression is up-regulated in ovarian cancer cells and may serve as a marker for ovarian cancer. Mucin-16 is shown to binds to mesothelin (MSLN) and this binding mediates heterotypic cell adhesion, which may contribute to the metastasis of ovarian cancer to the peritoneum by initiating cell attachment to the mesothelial epithelium.

Specificity

Clone 5E11 detects Mucin-16 in human ovarian cancer tissue. It targets an epitope within 264 amino acids from the C-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 264 amino acids from the C-terminal half of human Mucin-16.

Application

Anti-Mucin-16 (CA-125), clone 5E11, Cat. No. MABC1608, is a mouse monoclonal antibody that detects Mucin-16 and it has been tested for use in Immunohistochemistry (Paraffin), Proximity Ligation Assay, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected Mucin-16 in ovarian serous cystadenocarcinoma tissue (Courtesy of Dr. Leonor David, M.D., Ph.D., IPATIMUP and Medical Faculty of the University of Porto, Portugal).

Proximity Ligation Assay Analysis: A representative lot detected Mucin-16 in Proximity Ligation Assay applications (Ricardo, S., et. al. (2016). Virchows Arch. 468(6):715-22; Ricardo, S., et. al. (2015). Mol Oncol. 9(2):503-12).

Immunohistochemistry Analysis: A representative lot detected Mucin-16 in Immunohistochemistry applications (Marcos-Silva, L., et. al. (2015). Glycobiology. 25(11):1172-82; Ricardo, S., et. al. (2015). Mol Oncol. 9(2):503-12).

Immunohistochemistry Analysis: A representative lot detected Mucin-16 in ovarian serous cystadenocarcinoma tissue (Courtesy of Dr. Leonor David, M.D., Ph.D., IPATIMUP and Medical Faculty of the University of Porto, Portugal).

Western Blotting Analysis: A representative lot detected Mucin-16 in Western Blotting applications (Marcos-Silva, L., et. al. (2015). Glycobiology. 25(11):1172-82).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry in human ovarian cancer and human uterine tissues.

Immunohistochemistry (Paraffin) Analysis: A 1:50-250 dilution of this antibody detected Mucin-16 in human ovarian cancer and human uterine tissues.

Target description

1519.17 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service