Skip to Content
Merck

Skip To

MAB8564

Anti-Poliovirus 3 Antibody, clone 613-F1-B5-E5

ascites fluid, clone 613-F1-B5-E5, Chemicon®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
613-F1-B5-E5, monoclonal
Species reactivity:
human
Application:
IF
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

ascites fluid

antibody product type

primary antibodies

clone

613-F1-B5-E5, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

dry ice

Application

Anti-Poliovirus 3 Antibody, clone 613-F1-B5-E5 detects level of Poliovirus 3 & has been published & validated for use in IF.
IFA at 1:100-1:400.

Final working dilutions must be determined by end user.

SUGGESTED IFA

PROTOCOL:

Substrate slides:

1. Scrape and spot cells onto microscope slide wells.

2. Air dry.

3. Fix in cold acetone for 10 minutes at +4°C.

Stain:

1. Add 15 μL diluted polio antibody.

2. Incubate at +37°C for 30 minutes in a humid chamber.

3. Rinse in PBS for 5 minutes; repeat with fresh PBS.

4. Add 15 μL diluted labeled antibody.

5. Repeat steps 2 and 3.

6. Mount with buffered glycerol.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Biochem/physiol Actions

Reacts with the Poliovirus 3.

Preparation Note

Store at -20°C for up to 12 months in convenient, undiluted aliquots. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB8560MAB985MAB8256
clone

613-F1-B5-E5, monoclonal

clone

583-G8-G2-A4, monoclonal

clone

431-4H-6H-3B, monoclonal

clone

9B3.2, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

technique(s)

immunofluorescence: suitable

technique(s)

immunofluorescence: suitable

technique(s)

immunofluorescence: suitable

technique(s)

immunofluorescence: suitable

shipped in

dry ice

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters