Skip to Content
Merck
All Photos(1)

Key Documents

MAB3406

Sigma-Aldrich

Anti-Cytokeratin Pan Antibody, clone Lu5

clone Lu5, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

100
300

antibody form

purified antibody

antibody product type

primary antibodies

clone

Lu5, monoclonal

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG1

shipped in

wet ice

Specificity

The antibody reacts with an epitope which is common to all cytokeratins respectively with a cytokeratin-associated protein. The antigen recognized by anti-cytokeratin pan is present in all types of epithelia. The antibody reacts with epithelial and mesothelial cells. Exception: Secretory cells are not stained and acinus cells, for example, show only weak staining. The antibody does not react with connective tissue, blood or lymphatic vessels, smooth or skeletal muscles. Exception: When higher antibody concentrations are used, weakly positive reactions can be observed with the smooth muscles of the myometrium, the prostata as well as the gastric muscularis mucosa.The antibodies can be used for differential diagnosis of epithelial tumors, distinction from mesenchymal, neural tumors as well as large-cell lymphomas. Anti-cytokeratin pan stains all epithelial tumors (primary tumors as well as metastases). Exception: Only partially positive staining is observed with theca cell tumors, granulosa cell tumors, adrenal cortex tumors. In mixed tumors (e.g. carcinosarkoma, thymoma, adenolymphoma) only the epithelial regions are stained. Tumors of nonepithelial origin do not react with the antibody. Exception: With high antibody concentrations a weak staining of leiomyosarcoma of the uterus is observed.

Immunogen

Lung tumor cell lines A549 and A2182.

Application

Immunohistochemistry: 10-20 μg/mL Optimal working dilutions must be determined by end user.

Tumor Characterization

For tumor characterization a two-step experimental assay is recommended:

1. Characterization of the tissue origin of a tumor with the aid of anti-cytokeratin pan, anti-desmin, anti-glial fibrillary acidic protein, anti-neurofilament and anti-vimentin.

2. Characterization of an epithelial tumor using antibodies to cytokeratin classes which are characteristic for different tissues, e.g. anti-cytokeratin No. 7, anti-cytokeratin No. 18. Sample material: Normal tissue, tumor tissue following surgery or up to 24 h after autopsy, frozen sections or formaldehyde-fixed paraffin embedded sections,

Detection method

Immunohistochemical analysis (detection of the primary antibody e. g. by anti-mouse Ig-peroxidase or anti-mouse Ig-FITC).

Procedure:

Ideal frozen sections are obtained from shock-frozen tissue samples. The frozen sections are air-dried and then fixed with acetone for 5-10 min at -15 to -25°C. Excess acetone is allowed to evaporate at 15-25°C. Material fixed in alcohol can also be used.

Cytocentrifuge preparations of single cells or cell smears are also fixed in acetone. These preparations should, however, not be air dried, the excess acetone is removed by briefly washing in phosphate-buffered saline (PBS). Further treatment as follows:

• Overlay the preparation with 10 - 20 μL antibody solution and incubate in a humid chamber for 30 - 60 min at 15-25°C.

• Dip the slide briefly in PBS and then wash in PBS 3 times for 3 min each (using a fresh PBS bath in each case).

• Wipe the margins of the preparation dry, overlay the preparation with 10 - 20 μL of a solution of anti-mouse lg-FITC or anti-mouse Ig-peroxidase and allow to incubate in a humid chamber for 30 min at 15-25°C.

• Wash the slide in PBS as described above.

The preparation must not be allowed to dry out during any of the steps. lf using an indirect immunofluorescence technique, the preparation should be overlaid with a suitable embedding medium (e. g. Mowiol, Hoechst) and examined under the fluorescence microscope. lf a POD-conjugate is used as the secondary antibody, the preparation should be overlaid with a substrate solution (see below) and incubated at 15-25°C until a clearly visible redbrown color develops. A negative control (e.g. only the labeled secondary antibody) should remain unchanged in color during this incubation period.

Subsequently, the substrate is washed off with PBS and the preparation is stained with hemalum stain for about 1 min. The hemalum solution is washed off with PBS, the preparation is embedded and examined.

Substrate solutions:

Aminoethyl-carbazole: Dissolve 2 mg 3-amino-9-ethylcarbazole with 1.2 mL dimethylsulfoxide and add 28.8 mL Tris-HCI, 0.05 M; pH 7.3; and 20 μL 3% H 2 O 2 , (w/v). Prepare solution freshly each day. Diaminobenzidine: Dissolve 25 mg 3, 3′-diaminobenzidine with 50 mL Tris-HCI, 0.05 M; pH 7.3; and add 40 μL 3% H2O2 , (w/v). Prepare solution freshly each day. Note The antibody solutions have to be absolutely free of precipitate! lf necessary, centrifuge the solutions at high speed prior to use.
This Anti-Cytokeratin Pan Antibody, clone Lu5 is validated for use in IH for the detection of Cytokeratin Pan.

Physical form

Format: Purified
Liquid. Buffer = 0.02 M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

The lyophilized antibody is stabe when stored at 2-8°C.The reconstituted antibody solution is stable at 2-8°C for three monthes. The solution can be stored in aliquots at -15 to -25 °C for 12 monthes. Avoid repeat freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhairavi Bhatia et al.
Experimental eye research, 93(6), 852-861 (2011-10-13)
Much controversy has arisen on the nature and sources of stem cells in the adult human retina. Whilst ciliary epithelium has been thought to constitute a source of neural stem cells, a population of Müller glia in the neural retina
Z Zhang et al.
Neuroscience, 174, 10-25 (2010-12-01)
In adult cortices, the ratio of excitatory and inhibitory conductances (E/I ratio) is presumably balanced across a wide range of stimulus conditions. However, it is unknown how the E/I ratio is postnatally regulated, when the strength of synapses are rapidly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service