Skip to Content
Merck
All Photos(1)

Key Documents

MAB3310

Sigma-Aldrich

Anti-TIMP-2 Antibody, clone 67-4H11

clone 67-4H11, Chemicon®, from mouse

Synonym(s):

Tissue Inhibitor of Metalloproteinase-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

67-4H11, monoclonal

species reactivity

rabbit, goat, guinea pig, rat, human, bovine, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TIMP2(7077)

Specificity

The antibody specifically reacts with human TIMP-2.

SPECIES REACTIVITY:

Cross-reacts with mouse, rat, guinea pig, rabbit and bovine TIMP-2.

Immunogen

Synthetic peptide corresponding to amino acids 178-193 of human TIMP-2 (numbered from the propeptide)

YRGAAPPKQEFLDIED

Application

Anti-TIMP-2 Antibody, clone 67-4H11 detects level of TIMP-2 & has been published & validated for use in ELISA, WB, IH(P).
Immunoblotting

Immunohistochemistry on paraffin-embedded, PLP fixed tissue at 5 μg/mL (see Tomita reference) or frozen tissues at 1 μg/mL (see Ohashi reference).

ElA
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs

Linkage

Replaces: MAB3356

Physical form

Format: Purified
Liquid in 0.1M Na-Phosphate buffer, pH 7.0 containing 2% protease free bovine serum albumin.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Manufactured by Daiichi Fine Chemical Co., Ltd

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Galm et al.
Oncogene, 24(30), 4799-4805 (2005-05-05)
The tissue inhibitor of metalloproteinases-2 (TIMP-2) is known to antagonize matrix metalloproteinase activity and to suppress tumor growth, angiogenesis, invasion and metastasis. We analysed the methylation status of the CpG island in the TIMP-2 promoter region by methylation-specific polymerase chain
Wenni Tong et al.
Developmental neurobiology, 70(3), 182-194 (2009-12-18)
A recent study has shown that increased activity of matrix metalloproteinases-2 and metalloproteinases-9 (MMP-2 and MMP-9) has detrimental effect on the brain after neonatal hypoxia. The present study determined the effect of maternal hypoxia on neuronal survivability and the activity
TNF-alpha stimulates activation of pro-MMP2 in human skin through NF-(kappa)B mediated induction of MT1-MMP.
Han, YP; Tuan, TL; Wu, H; Hughes, M; Garner, WL
Journal of Cell Science null
Christina Tekle et al.
International journal of cancer, 130(10), 2282-2290 (2011-06-15)
B7-H3, an immunoregulatory protein, is known to play a role in tumor progression. In many cancer types, observed correlations between high B7-H3 expression and poor prognosis have been attributed to involvement in antitumor immunity. However, here we demonstrate a nonimmunological
Tao Xu et al.
Experimental and therapeutic medicine, 10(2), 683-688 (2015-12-02)
The mortality rates associated with colorectal cancer (CRC) are high due to metastasis. Epithelial-to-mesenchymal transition (EMT) is a key step in tumor metastasis. The aim of the present study was to investigate the function of microRNA-20a (miR-20a) in EMT. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service