Skip to Content
Merck
All Photos(1)

Documents

MAB1969-I

Sigma-Aldrich

Anti-Integrin α5β1 Antibody

Synonym(s):

Integrin antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

JBS5, monoclonal

mol wt

observed mol wt ~114.54 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
flow cytometry: suitable
inhibition assay: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Integrin alpha-5 beta1 (UniProt: P08648/P05556); also known as CD49 antigen-like family member E, Fibronectin receptor subunit alpha/Fibronectin receptor subunit beta, VLA-5/ VLA-4 subunit beta, CD49e/CD29) is encoded by the ITGA5/ITGB1 (also known as FNRA/FNRB, MDF2, MSK12) gene (Gene ID: 3678/3688) in human. Integrins are heterodimeric integral membrane proteins composed of an a subunit and a b subunit that function in cell surface adhesion and signaling. Integrin a5 is expressed on thymocytes, T cells, monocytes, platelets, early B cells, and activated in B cells. Integrin a5 exhibits an increased expression in mesodermal progenitors and is shown to be prevalent in fetal cardiomyocytes. Its deletion is shown to cause mesodermal defects during embryogenesis in murine models. Membrane fractions obtained from fibroblasts of diabetic animals display significant reduction in integrin a5 levels. Integrin a5 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-41), which is subsequently cleaved off to generate the mature form that contains a heavy chain (aa 42-894) and a light chain (aa 895-1049). Structurally, it consists of an extracellular domain (aa 42-995), a transmembrane domain (aa 996-1021), and a cytoplasmic domain (aa 1022-1049). It contains four calcium binding sites (aa 480-488; 334-342; 401-409; and 465-473). When associated with b 1 subunit it can serve as a receptor for fibronectin and fibrinogen. It recognizes the sequence R-G-D in its ligands. Integrin b 1 is synthesized with a signal peptide (aa 1-20) that is subsequently cleaved off. The mature form has an extracellular domain (aa 21-728), a helical domain (aa 729-751) and a cytoplasmic region (aa 752-798). (Ref.: Almeida, ME et al., (2016). Mol. Cell. Biochem. 421(1-2); 19-28; Neiman, G., et al. (2019). Sci. Rep. 9; 18077; Su, Y., et al. (2016). Proc. Natl. Acad. Sci. USA. 113(27); E3872-E3881).

Specificity

Clone JBS5 is a mouse monoclonal antibody that detects human Integrin a5b1. It targets and epitope within the extracellular domain.

Immunogen

Jurkat cells.

Application

Quality Control Testing

Evaluated by Flow Cytometry in HT-1080 cells.

Flow Cytometry Analysis: 1 μg of this antibody detected Integrin a5b1 in one million HT-1080 cells.

Tested Applications

ELISA Analysis: A representative lot detected Integrin a5 in ELISA applications (Mould, A.P., et. al. (1998). Biochem J. 331 ( Pt 3)(Pt 3):821-8).

Western Blotting Analysis: A representative lot detected Integrin a5 in Western Blotting applications (Ria, R., et. al. (2002). Haematologica. 87(8):836-45; Gu, C., et. al. (2001). Mol. Cell. Biol. 21(14):4579-97).

Immunocytochemistry Analysis: A representative lot detected Integrin a5 in Immunocytochemistry applications (Gong, X., et. al. (2018). EMBO J. 37(17):e98994).

Inhibition Analysis: A representative lot of this antibody produced inhibition of fibrinogen-, fibronectin- and laminin-mediated cell adhesion. (Wattam, B., et. al. (2001). Biochem J. 356(Pt 1):11-7; Rahman, S., et. al. (1998). Biochem J. 335 (Pt 2)(Pt2):247-57; Mould, A.P., et. al. (1998). Biochem J. 331 ( Pt 3)(Pt 3):821-8; Gu, C., et. al. (2001). Mol Cell Biol. 21(14):4579-97).

Immunohistochemistry Applications: A representative lot detected Integrin 5?1 in Immunohistochemistry applications (Hernandez, P.A., et. al. (2020). Sci Rep. 10(1):627; Qin, L., et. al. (2003). Biol Reprod. 69(2):563-71).

Immunoprecipitation Analysis: A representative lot detected Integrin 5?1 in Immunoprecipitation applications (Nishida, T., et. al. (2007). J Cell Commun Signal. 1(1):45-58; Gu, C., et. al. (2001). Mol Cell Biol. 21(14):4579-97).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Integrin a5β1, clone JBS5, Cat. No. MAB1969-I, is a mouse monoclonal antibody that detects Integrin a5b1 and is tested in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Inhibition Assay, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service