Skip to Content
Merck
All Photos(1)

Key Documents

MAB1678

Sigma-Aldrich

Anti-Filamin A Antibody, clone PM6/317

ascites fluid, clone PM6/317, Chemicon®

Synonym(s):

Alpha-Filamin, Filamin I, Endothelial Actin-binding Protein, ABP-280, Nonmuscle Filamin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PM6/317, monoclonal

species reactivity

human, rabbit, rat, guinea pig, chicken, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FLNA(2316)
mouse ... Flna(192176)

General description

Filamin is a structural protein that forms flexible cross-links between two actin filaments. Filamin is a homodimer of polypeptide chains each joined to the other at one end with an actin binding site ath the other. It is present in smooth muscle, fibroblasts, platelets and lymphocytes.

Specificity

Expected to cross-react with chicken, guinea pig and rabbit.
Recognizes unprocessed, full length Human filamin (actin-binding protein; 270-280 kDa) as well as the 190 kDa N-terminal calpain cleavage fragment of filamin (Aakhus, 1992). Following induction of apoptosis in U937 cells, the antibody recognizes 170, 150, and 120 kDa N-terminal cleavage fragments of the full-length form presumably resulting from cleavage by activated caspase-3 (Umeda, 2001).

Immunogen

Human platelet protein.

Application

Immunoblotting:
1:1000-1:4000. Because of the large size of the unprocessed forms of filamin, 4-7% PAGE gels and proteinase inhibitors are recommended.

Immunofluorescence:
1:50-1:200 dilution from a previous lot was used. Suitable for staining both frozen and paraffin embedded tissues (at lower dilutions). Microwave-citrate buffer antigen retrieval method recommended for paraffin sections.

Immunoprecipitation:
A previous lot was used on immunoprecipitation. Suggested lysis buffer is PBS with 0.5% triton X-100 with proteinase inhibitors (note for full length filamin include calpain inhibitors). 5 microliters of antibody for every 300-500 μL of cell lysate (200-500 μg/mL total protein is suggested. Incubation is 1 hour RT or overnight 4C; Protein A/G agarose beads or rabbit anti-mouse secondary capture antibody is recommended for best recovery. 4-8% acrylamide gels are recommended for full length filamin or the 190 kDa fragement visualization.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Filamin A Antibody, clone PM6/317 is validated for use in IF, IH, IH(P), IP, WB for the detection of Filamin A.

Quality

Routinely evaluated by Western Blot on Jurkat lysates.

Western Blot Analysis:
1:500-1:4000 dilution of this lot detected Filamin A on 10 μg of Jurkat lysates. Because of the large size of the unprocessed forms of filamin, 4-7% PAGE gels and proteinase inhibitors are recommended.

Target description

270-280 kDa & 190 kDa

Physical form

Mouse monoclonal ascites IgG1 in buffer containing 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquots from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive control tisse: skin, jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ribosomal S6 kinase (RSK) regulates phosphorylation of filamin A on an important regulatory site.
Woo, MS; Ohta, Y; Rabinovitz, I; Stossel, TP; Blenis, J
Molecular and cellular biology null
Qun Lu et al.
Journal of Alzheimer's disease : JAD, 22(1), 235-245 (2010-09-18)
Presenilin mutations are linked to the early onset familial Alzheimer's disease (FAD) and lead to a range of neuronal changes, indicating that presenilins interact with multiple cellular pathways to regulate neuronal functions. In this report, we demonstrate the effects of
Eleonora Vitali et al.
Endocrine-related cancer, 23(3), 181-190 (2016-01-07)
Somatostatin receptor type 2 (SST2) is the main pharmacological target of somatostatin (SS) analogues widely used in patients with pancreatic neuroendocrine tumours (P-NETs), this treatment being ineffective in a subset of patients. Since it has been demonstrated that Filamin A
Xi Jiang et al.
International journal of biological sciences, 9(1), 67-77 (2013-01-05)
Filamin-A cross-links actin filaments into dynamic orthogonal networks, and interacts with an array of proteins of diverse cellular functions. Because several filamin-A interaction partners are implicated in signaling of cell mobility regulation, we tested the hypothesis that filamin-A plays a
L Valmu et al.
European journal of immunology, 29(7), 2107-2118 (1999-07-31)
Leukocyte adhesion is a regulated process, which involves CD11/CD18 leukocyte integrins. CD11/CD18 acidity may be regulated intracellularly, and the CD18 polypeptide has previously been shown to become phosphorylated on serine and threonine after phorbol ester activation of T cells. Increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service