MAB1535
Anti-Hexokinase Type III Antibody, clone C7C3
ascites fluid, clone C7C3, Chemicon®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
C7C3, monoclonal
species reactivity
rat
manufacturer/tradename
Chemicon®
technique(s)
immunocytochemistry: suitable
isotype
IgG1
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
rat ... Hk1(25058)
Specificity
Recognizes Type III hexokinase. No cross-reactivity with Type I or Type II isozymes.
Immunogen
Rat Type III hexokinase.
Application
Anti-Hexokinase Type III Antibody, clone C7C3 detects level of Hexokinase Type III & has been published & validated for use in IC.
Immunohistochemical localization of Type III hexokinase: 1:50.
Optimal working dilutions must be determined by end user.
Optimal working dilutions must be determined by end user.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service