Skip to Content
Merck
All Photos(6)

Key Documents

MAB1281

Sigma-Aldrich

Anti-Nuclei Antibody, clone 235-1

clone 235-1, Chemicon®, from mouse

Synonym(s):

HuNu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
235-1, monoclonal
application:
ICC
IHC
IP
species reactivity:
human
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
citations:
362

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

235-1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable

input

sample type neural stem cell(s)

isotype

IgG1

shipped in

dry ice

target post-translational modification

unmodified

General description

Cell replacement techniques such as stem-cell or fetal-cell therapies show great promise for many diseases. Cross-species grafting between human and animal cells or tissues requires the identification and localization of the donor material in the host environment. Anti-nuclei antibodies are useful cytological and histological markers for identifying human cells in xenograft models.

Specificity

Stains nuclei of all human cell types giving a diffuse nuclear staining pattern. Chromosomes are negatively stained in metaphase cells. Also stains nuclei from primate (Sanchez-Pernaute, 2005). The antibody does not react with nuclei from mouse or rat.
The human nuclei monoclonal reacts specifically with human cells. It shows no reactivity against mouse, rat, chicken in immunohistorchemistry. Other mammalian or animal species have not been tested for reactivity.

Application

Anti-Nuclei Antibody, clone 235-1 has been used in: Immunohistochemistry (1:200); Immunolabeling for human nuclei (HuNu) (1:300); Histological and immuno-fluorescent analysis of submandibular glands

Physical form

Format: Purified
Hybridoma supernatant. Liquid in PBS. Contains no preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
All human cell types

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Lei et al.
American journal of obstetrics and gynecology, 212(5), 639-639 (2015-01-04)
Using a mouse model of intrauterine inflammation, we have demonstrated that exposure to inflammation induces preterm birth and perinatal brain injury. Mesenchymal stem cells (MSCs) have been shown to exhibit immunomodulatory effects in many inflammatory conditions. We hypothesized that treatment
Yuelin Zhang et al.
Cell transplantation, 21(10), 2225-2239 (2012-07-11)
Although transplantation of adult bone marrow mesenchymal stem cells (BM-MSCs) holds promise in the treatment for pulmonary arterial hypertension (PAH), the poor survival and differentiation potential of adult BM-MSCs have limited their therapeutic efficiency. Here, we compared the therapeutic efficacy
E Josue Ruiz et al.
Nature communications, 5, 3175-3175 (2014-01-17)
The signals that regulate stem cell self-renewal and differentiation in the lung remain elusive. Lung stem cells undergo self-renewal or lineage commitment to replenish tissue, depending on cross-talk with their environment. This environment, also known as the niche, includes mesenchymal
Human stem cell delivery for treatment of large segmental bone defects.
Dupont, KM; Sharma, K; Stevens, HY; Boerckel, JD; Garcia, AJ; Guldberg, RE
Proceedings of the National Academy of Sciences of the USA null
Ji Hyun Kim et al.
Scientific reports, 6, 38754-38754 (2016-12-08)
Reconstructing functional volumetric tissue in vivo following implantation remains a critical challenge facing cell-based approaches. Several pre-vascularization approaches have been developed to increase cell viability following implantation. Structural and functional restoration was achieved in a preclinical rodent tissue defect; however

Articles

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Learn differences in monoclonal vs polyclonal antibodies including how antibodies are generated, clone numbers, and antibody formats.

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

See All

Protocols

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service