M96ULA
Millicell® Ultra-low Attachment Plates
96-well, ultra-hydrophilic polymer coated, u-bottom, skirt, polystyrene, 20 pack, Spheroid and Organoid Culture
Synonym(s):
96-well ULA Plates, ULA Plates
About This Item
Recommended Products
material
polystyrene (ultra-hydrophilic polymer coated)
round clear plastic bottom
Quality Level
description
ultra-hydrophilic polymer coated, u-bottom
feature
skirt
specific plates for 3D cell culture
packaging
pack of 20
manufacturer/tradename
Millicell®
well size
96 wells
well working volume
300 μL , 250UL recommended
General description
Application
Features and Benefits
- Non-binding surface for cells to facilitate natural spheroid formation
- Uniform single spheroid/EB formation in each well
- Spheroid assay formation and analysis in the same plate
- High optical clarity plates for imaging
- Stable, non-cytotoxic and cell non-adhesion surface
- Easy handling, compatible with liquid robotic system
- Suitable for brightfield imaging and fluorescence microscopy
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn how to grow spheroid cultures in ULA plates, the common applications and benefits of ULA plates for 3D cell culture, Millicell® ULA plate dimensions, plate handling, troubleshooting, and more.
Learn about the similarities and differences in spheroid formation, roundness, and circularity between Millicell® ultra-low attachment plates and other ULA plates.
Learn how to quantify spheroid formation, roundness, and circularity. Discover Millicell® Ultra-low Attachment plates, an ideal 3D cell culture model for uniform spheroid formation.
Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.
Protocols
Discover our protocol for thawing, plating, and forming hepatocyte spheroids from human spheroid-qualified plateable hepatocytes. Find the formulation for hepatocyte spheroid maintenance media.
Related Content
Tools to accelerate preclinical studies in antibody drug development, from toxicity to immunogenicity, pharmacokinetics, pharmacodynamics, and efficacy.
Explore our preclinical models, tools for 2D and 3D cell culture, cell-based assays, reagents, instruments, and more to support antibody drug development.
Millicell® Ultra-low Attachment plates facilitate 3D cell culture with detailed application information for cell seeding to analysis.
Explore our portfolio of 3D cell culture tools and technologies for any 3D cell line or application and discover organoids, hydrogels, ULA, microwell, or hydrogel plates, and more.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service