Skip to Content
Merck
All Photos(2)

Documents

LSKMAGN01

Millipore

PureProteome NHS FlexiBind Magnetic Beads Kit

NHS FlexiBind beads are suitable for Immunoprecipitations, purifying nucleic acids, isolating cells and organelles, performing protein-protein interaction studies and many other applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
eCl@ss:
32160405
NACRES:
NA.56

packaging

kit of 0.5 mL beads

manufacturer/tradename

PureProteome

technique(s)

protein purification: suitable

particle size

10 μm

capacity

>17 μmol/mL, settled beads binding capacity (NHS)

shipped in

wet ice

storage temp.

2-8°C

General description

PureProteome NHS (N-Hydroxysuccinimide) FlexiBind Magnetic Beads provide researchers flexibility in binding the ligand of their choice. The only prerequisite is that the molecule must contain a primary free Amine group. NHS FlexiBind beads are suitable for Immunoprecipitations, purifying nucleic acids, isolating cells and organelles, performing protein-protein interaction studies and many other applications.

Application

Research Category
All

Components

0.5 mL NHS magnetic beads, 20% slurry, 4 Pk Amicon® Ultra-0.5 Centrifugal Filters, 30,000 MWCO, 5 mL Equilibration Buffer, 25 mL Coupling & Wash Buffer, 5 mL Quenching Buffer

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Darryl A Wesener et al.
eLife, 10 (2021-03-09)
Methods for measuring gut microbiota biochemical activities in vivo are needed to characterize its functional states in health and disease. To illustrate one approach, an arabinan-containing polysaccharide was isolated from pea fiber, its structure defined, and forward genetic and proteomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service