L25DET6S1
Millistak+® Lenticular Disc Filters, DE25 media series, 16 in. diameter, 16-cell, silicone seal
About This Item
material
cellulose
polypropylene
silicone seal
Quality Level
description
sanitization May be hot water sanitized for 20 cycles using purified water @ 80 °C for 30 minutes
sterility
(May be steam sterilized for 3 steam-in-place (SIP) cycles of 30 minutes @ 123 °C)
feature
hydrophilic
conductivity measuring range
9-12 μmho/cm
parameter
0.03 bar max. differential pressure (0.5 psid) (Reverse)
0.2 bar max. inlet pressure (3 psi) at 123 °C
0.21 bar max. differential pressure (3 psid) at 123 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
1.03 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.07 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. inlet pressure (30 psi) at 25 °C
H
27.6 cm (10.88 in.)
diam.
16 in. , 16-cell
40.64 cm (16 in.)
filtration area
3.47 m2
impurities
<0.25 EU/mL Aqueous extraction (LAL test)
total organic carbon (TOC) residue
753-1567 ppb
filter grade
DE25
General description
Analysis Note
NVR: 3.93–6.43 mg/m2
TOC: 753–1,567 ppb
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service