Skip to Content
Merck
All Photos(1)

Documents

IPVH07850

Millipore

Immobilon® -P PVDF Membrane

50 sheets, 7 cm x 8.4 cm, 0.45 µm pore size, transfer membrane

Synonym(s):

Western blotting membrane, blotting membrane, mini blots, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

product name

Immobilon®-P PVDF Membrane, 50 sheets, 7 cm x 8.4 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

7 cm × 8.4 cm

pore size

0.45 μm pore size

capacity

160 μg/cm2 adsorption capacity (insulin)
215 μg/cm2 adsorption capacity (BSA)
294 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Toluidine blue
for use with Transillumination

detection method

chemiluminescent
colorimetric
fluorometric
radioactive

shipped in

ambient

Related Categories

General description

Immobilon®-P transfer membrane is a polyvinylidene fluoride (PVDF) microporous membrane with a nominal pore size of 0.45 micron (μm). It is recommended for western blotting, especially for proteins greater than 20 kDa. Immobilon®-P transfer membrane can bind a wide range of molecular weight proteins. It is used for the transfer of proteins from a variety of gel matrices. When compared to a nitrocellulose membrane, the Immobilon-P PVDF membrane has a higher signal-to-noise ratio for enhanced sensitivities, improved handling characteristics and staining capabilities and increased solvent resistance.
Primary Binding Mechanisms: Electrostatic, hydrophobic;
Filter Code: IPVH

Application

Immobilon®-P PVDF Membrane has been used in western blotting.

Features and Benefits

Immobilon®-P membrane has:
  • excellent protein retention
  • high physical strength
  • broad chemical compatibility
  • suitable for a variety of staining applications and reprobing in immunodetection methods

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Kang et al.
American journal of otolaryngology, 33(6), 678-684 (2012-07-10)
The purposes of this study were to determine whether autophagy was involved in cisplatin (CDDP) resistance and to investigate the role of the autophagy in the regulation of chemosensitivity to CDDP in laryngeal cancer Hep-2 cells. A WST-1 assay was
Paola Maycotte et al.
Autophagy, 8(2), 200-212 (2012-01-19)
Chloroquine (CQ) is a 4-aminoquinoline drug used for the treatment of diverse diseases. It inhibits lysosomal acidification and therefore prevents autophagy by blocking autophagosome fusion and degradation. In cancer treatment, CQ is often used in combination with chemotherapeutic drugs and
Sheikh F Ahmad et al.
Neurotoxicology, 77, 1-11 (2019-12-08)
Autism spectrum disorder (ASD) comprises a broad range of neurodevelopmental disorders that are associated with deficits in social interaction and communication. The tyrosine kinase inhibitor tyrphostin AG126 represents a promising therapeutic agent for several neuroinflammatory disorders. There are currently no
Kentaro Togashi et al.
Journal of the neurological sciences, 413, 116802-116802 (2020-04-04)
Parkinson's disease (PD) is characterized by progressive degeneration of dopaminergic neurons in the substantia nigra pars compacta (SNc). Levodopa (L-Dopa), the current main treatment for PD, reduces PD symptoms by partially replacing dopamine, but it does not slow neurodegeneration. Recent

Protocols

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service