HX0290
Hexane, mixture of isomers
≥98.5% (total hexane isomers + methylcyclopentane), suitable for HPLC, meets USP/NF
Synonym(s):
Hexanes, FM approval
About This Item
Recommended Products
Product Name
Hexanes, HPLC, 98.5% Hexanes, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Agency
meets USP/NF
Quality Level
vapor pressure
160 hPa ( 20 °C)
Assay
≥98.5% (total hexane isomers + methylcyclopentane)
form
liquid
technique(s)
HPLC: suitable
impurities
≤0.0003 meq/g Water-soluble titrable acid
≤0.005% Sulfur compounds (as S)
≤0.01% Water (H2O)
evapn. residue
≤5 ppm
color
APHA: ≤10
bp
69 °C/1013 hPa
mp
-94.3 °C
transition temp
flash point -22 °C
solubility
0.0095 g/L
density
0.66 g/cm3 at 20 °C
UV absorption
λ: 195 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.30
λ: 220 nm Amax: ≤0.20
λ: 230 nm Amax: ≤0.10
λ: 240 nm Amax: ≤0.04
λ: 250 nm Amax: ≤0.02
λ: 280-400 nm Amax: ≤0.01
shipped in
ambient
storage temp.
room temp
SMILES string
C(CCC)CC
InChI
1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
InChI key
VLKZOEOYAKHREP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
HX0290-NP10 sold as 2 x 10 L
Preparation Note
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system, Nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-7.6 °F - closed cup
Flash Point(C)
-22 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service