Skip to Content
Merck
All Photos(2)

Key Documents

HCP2MAG-62K-PX23

Millipore

MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel II - Premixed 23 Plex - Immunology Multiplex Assay

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 81-110%
sensitivity: 0.4-55.8 pg/mL
standard curve range: 1.0-100,000 pg/mL

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

“Cytokine” is a general term used for a diverse group of soluble proteins and peptides which act as regulators under both normal and pathological conditions to modulate the functional activities of individual cells and tissues. These proteins also mediate direct interactions between cells and regulate processes taking place in the extracellular environment. The cytokine group of proteins includes lymphokines, interferons, colony stimulating factors and chemokines. Cytokine and chemokine research plays a significant role in achieving a deeper understanding of the immune system and its multi-faceted response to most antigens, as well as disease states such as inflammatory disease, allergic reactions, irritable bowel disease (IBD), sepsis, and cancer.

The MILLIPLEX® Human Cytokine / Chemokine Panel II enables you to focus on the therapeutic potential of cytokines as well as the modulation of cytokine expression.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There was no or negligible cross-reactivity between the antibodies and any of the other analytes in this panel.

Application

  • Analytes: 6Ckine, BCA-1, CTACK, ENA-78, Eotaxin-2, Eotaxin-3, I-309, IL-16, IL-20, IL-21, IL-23, IL-28A, IL-33, LIF, MCP-2, MCP-4, MIP-1δ, SCF, SDF-1A+β, TARC, TPO, TRAIL, TSLP
  • Recommended Sample type: Serum, plasma, cell culture supernatant
  • Recommended Sample dilution: neat
  • Assay Run Time: Overnight or two-hour primary incubation. For best results, an overnight incubation is recommended
  • Research Category: Inflammation & Immunology
  • Research Subcategory: Obesity, Metabolic Disorders, Inflammation & Autoimmune Mechanisms

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Please contact Technical Service for linearity of dilution.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holly Mansell et al.
American journal of nephrology, 41(2), 147-155 (2015-04-01)
The 7-year Major Adverse Cardiovascular Events Calculator (CRCRTR-MACE) predicts cardiovascular events (CVE) in renal transplant recipients (RTR), and thrombopoietin (TPO) is a humoral inflammatory factor implicated in cardiovascular disease (CVD). The aim of the study was to determine if circulating
Gerald V Denis et al.
PloS one, 13(5), e0196755-e0196755 (2018-05-09)
Obesity-driven Type 2 diabetes (T2D) is a systemic inflammatory condition associated with cardiovascular disease. However, plasma cytokines and tissue inflammation that discriminate T2D risk in African American women with obese phenotypes are not well understood. We analyzed 64 circulating cytokines
Marion C Lanteri et al.
The Journal of infectious diseases, 211(10), 1585-1596 (2014-11-27)
An association between GB virus C (GBV-C) and improved outcomes of human immunodeficiency virus (HIV) infection has been reported in HIV-positive individuals with active GBV-C coinfection. This study provides insights into the immune mechanisms underlying the protective role of GBV-C
Sarah M Engle et al.
Clinical and experimental immunology (2022-01-13)
The pathogenesis of atopic dermatitis (AD) results from complex interactions between environmental factors, barrier defects, and immune dysregulation resulting in systemic inflammation. Therefore, we sought to characterize circulating inflammatory profiles in pediatric AD patients and identify potential signaling nodes which
Susan Louise Lindsay et al.
Stem cell reports, 6(5), 729-742 (2016-04-28)
Previously we reported that nestin-positive human mesenchymal stromal cells (MSCs) derived from the olfactory mucosa (OM) enhanced CNS myelination in vitro to a greater extent than bone-marrow-derived MSCs (BM-MSCs). miRNA-based fingerprinting revealed the two MSCs were 64% homologous, with 26 miRNAs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service