HAWG0470
MCE Membrane Filter, 0.45 μm Pore Size
MF-Millipore™, filter diam. 47 mm, hydrophilic, white, gridded, pkg of 500 discs, thickness 150 μm
Synonym(s):
MF-Millipore™ Membrane Filter, 0.45 µm pore size, gridded
About This Item
Recommended Products
material
gridded
mixed cellulose esters (MCE) membrane
white filter
Quality Level
description
47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 500 discs
sterility
non-sterile
feature
hydrophilic
packaging
pkg of 500 discs
manufacturer/tradename
MF-Millipore™
Millipore
parameter
4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.
diam.
47 mm
filter diam.
47 mm
thickness
150 μm
gravimetric extractables
2.5%
color
white
refractive index
n/D 1.51
matrix
MF-Millipore™
pore size
0.45 μm pore size
79 % porosity
bubble point
≥1.8 bar, air with water at 23 °C
Related Categories
General description
MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Application
- Clarification of aqueous solutions
- Particle removal
- Particle analysis
- Microbiology analysis
- Isolation of virus-like particles in wastewater
- Microplastics analysis grade water
- Nucleic acid binding, including eDNA
Other Notes
- Organism Retention: Microorganism
- Mode of Action: Filtration (size exclusion)
- Application: General laboratory filtration
- Intended Use: Retention or removal of biological contaminants
- Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
- Storage Statement: Store in dry location away from heat source
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service