Skip to Content
Merck
All Photos(1)

Key Documents

HABG047S0

Millipore

SO-Pak Filters 0.45µm 47mm black gridded

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
pore size:
0.45 μm pore size
material:
black filter
gridded filter
mixed cellulose esters (MCE)
feature:
hydrophilic
filter diam.:
47 mm
sterility:
sterile; ethylene oxide treated
packaging:
box of 10 envelopes containing 10 pak

material

black filter
gridded filter
mixed cellulose esters (MCE)

sterility

sterile; ethylene oxide treated

feature

hydrophilic

packaging

box of 10 envelopes containing 10 pak

filter diam.

47 mm

pore size

0.45 μm pore size

input

sample type cider
sample type food(s)
sample(s) (environmental)
sample type pharmaceutical(s)
sample type sport drink
sample type beer
sample type soft drink
sample type cosmetics
sample type beverage(s)
sample type wine
sample type bottled water

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

shipped in

ambient

General description

SO Pak filters - 10 resealable envelopes containing 10 filters and 10 pads, presterilized.

Application

Microbiological analysis

Packaging

Sterile pack: 10 resealable envelopes containing 10 filters and 10 pads, sterile

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service