Skip to Content
Merck
All Photos(1)

Key Documents

GK-1L-BK

Millipore

Anti-e (clone MS-62/MS-69) Blood Grouping Reagent

BIOSCOT Anti-e from the clone MS-62/MS-69, Human IgM, Blood Grouping Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

Quality Level

antibody form

unpurified

clone

MS-62/MS-69, monoclonal

form

liquid

packaging

pkg of 1 L

manufacturer/tradename

BIOSCOT

concentration

20-41 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.0-7.4

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 30 months from date of filtration

Product formulation examined according to Annex IV of the EU IVD directive to support CE registration.

For In Vitro use only

Application

Bulk
For use by Tube;Slide;Microplate;Gel and Column techniques

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service