SureWick® Pad Materials are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. SureWick® glass fiber conjugate / diagnostic pads are used to release detector reagents and faciltiate sample flow across the lateral flow membrane. This pack contains 100 strips of 8mm x 300mm glass fiber diagnostic pad (GFDX) material.
Application
SureWick® Pad Materials are optimised for use as sample, conjugate, or absorbent pads in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.
Features and Benefits
SureWick® glass fiber pads have low extractables and excellent consistency.
They provide reliable, predictable sample flow and reagent delivery.
They exhibit consistent thickness, bed volume, and uniform surface quality.
Quick Delivery and Lead time.
Excellent Technical support.
Legal Information
SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany