Skip to Content
Merck
All Photos(2)

Key Documents

FCMAB113A4

Sigma-Aldrich

Anti-Oct-4 Antibody, clone 10H11.2, Alexa Fluor 488 conjugate

clone 10H11.2, from mouse, ALEXA FLUOR 488

Synonym(s):

Octamer-binding transcription factor 3, POU class 5 homeobox 1, POU domain class 5, transcription factor 1, POU domain, class 5, transcription factor 1, POU-type homeodomain-containing DNA-binding protein, octamer-binding transcription factor-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

10H11.2, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POU5F1(5460)

Related Categories

General description

Octamer-4 (Oct-4), a member of the POU family of transcription factors, has been demonstrated to be vital for the formation of self-renewing pluripotent stem cells. During embryogenesis expression of Oct-4 is limited to pluripotent cells of the inner cell mass (ICM) that contribute to the formation of all fetal cell types. This relationship between Oct-4 and pluripotency has seen this transcription factor emerge as a marker of pluripotent stem cells. Undifferentiated human and murine pluripotent embryonic stem (ES) and embryonic carcinoma (EC) cells express Oct-4. Additionally, murine embryonic germ (EG) cells are also known to express Oct-4. Following stem cell differentiation, the level of Oct-4 expression decreases. Oct-4 expression by human EG cells is not known.

Specificity

Reacts with human Oct-4.

Immunogen

Recombinant GST fusion protein, human Oct-4 (aa 1-134).

Application

Detect Oct-4 using this Anti-Oct-4 Antibody, clone 10H11.2, Alexa Fluor 488 conjugate validated for use in FC & IC.
Flow Cytometry:
Antibody dilution for cellular staining:
• Prepare an antibody working solution by diluting 1:5 the primary antibody with PBS.
• Dispense the volume per test of working of solution according to the number of cells indicated in the table below.
• 5 µL for Guava Flow Cytometer
• 10 µL for other Flow Cytometry instruments
Working Solution Cells / test Total Reaction Volume
5 μl 5 x 105 Cells / 95 μl PBS 100 μl
10 μl 1 x 106 Cells / 90 μl PBS 100 μl
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Flow Cytometry in 2102Ep Cells.

Target description

~44 kDa

Physical form

Purified mouse monoclonal IgG1 conjugated to Alexa Flour 488 in PBS with less than 0.09% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt

Analysis Note

Control
Human embryonic stem cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaohui Pan et al.
PloS one, 10(1), e0114423-e0114423 (2015-01-21)
Undifferentiated stem cells are better donor cells for somatic cell nuclear transfer (SCNT), resulting in more offspring than more differentiated cells. While various stem cell populations have been confirmed to exist in the skin, progress has been restricted due to
Patricia Ablanedo Morales et al.
Biofabrication, 15(3) (2023-05-10)
Since the first description of inkjet bioprinting of cells in 2003, quantifying the input and measuring the output of the printers has been the hallmark of the field of bioprinting, as it is virtually impossible to characterize cells that are
Jeanette Beers et al.
Nature protocols, 7(11), 2029-2040 (2012-10-27)
This protocol describes an EDTA-based passaging procedure to be used with chemically defined E8 medium that serves as a tool for basic and translational research into human pluripotent stem cells (PSCs). In this protocol, passaging one six-well or 10-cm plate
David G Belair et al.
Scientific reports, 10(1), 2864-2864 (2020-02-20)
Exposure to thalidomide during a critical window of development results in limb defects in humans and non-human primates while mice and rats are refractory to these effects. Thalidomide-induced teratogenicity is dependent on its binding to cereblon (CRBN), the substrate receptor
Vivek Shukla et al.
Cancer research, 77(22), 6267-6281 (2017-09-25)
In this study, we generated induced pluripotent stem cells (iPSC) from normal human small airway epithelial cells (SAEC) to investigate epigenetic mechanisms of stemness and pluripotency in lung cancers. We documented key hallmarks of reprogramming in lung iPSCs (Lu-iPSC) that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service