Skip to Content
Merck
All Photos(1)

Documents

EZHS42

Millipore

Human Amyloid β42 ELISA Kit

measures and quantifies Amyloid β42 levels in 50 μL CSF, cell culture supernatent or plasma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

product name

High Sensitivity Human Amyloid β42 ELISA, This High Sensitivity Human Amyloid β42 ELISA is used to measure & quantify Amyloid β42 levels in Neuroscience research.

Quality Level

species reactivity

human

packaging

kit of 1 × 96 wells

parameter

50 μL sample volume (Overnight assay)

assay range

sensitivity: 8.0 pg/mL
(50 μl sample size)

standard curve range: 16-500 pg/mL

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum
sample type cerebrospinal fluid (CSF)

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... APP(351)

General description

Amyloid beta peptides have been implicated in the etiology of Alzheimer’s disease. Amyloid beta 40 is the most prominent peptide and Amyloid beta 42 is the neurotoxic form. The Amyloid beta 42/40-ratio (AB ratio) has been reported as a better indicator of the Alzheimer pathology. Millipore’s High Sensitivity Human Amyloid β42 ELISA kit is used for the measurement of Amyloid β42 in cerebrospinal fluid, cell culture supernatants, primary neurons and plasma in a 96-well format.

Specificity

The Amyloid β42 ELISA (HS) uses monoclonal anti-Aβ antibodies with high selectivity for human Aβ. The capture antibody recognizes the C-terminal end of Amyloid β1-42, which causes a high selectivity for Aβ42. The cross-reactivity of the used antibodies to other Amyloid peptides was tested by ELISA and BIACORE and shows no significant cross-reactivity to Aβ1-38, Aβ1-39, Aβ1-40, Aβ1-43 and Aβ1-44.

Application

Research Category
Neuroscience
Research Sub Category
Alzheimer′s Disease
This High Sensitivity Human Amyloid β42 ELISA is used to measure & quantify Amyloid β42 levels in Neuroscience research.
This assay requires 50 µl of sample and is an overnight assay.
Used to detect/quantify: Amyloid β42

Storage and Stability

Components in the kit can be stored up to 2 weeks at 2-8°C

Other Notes

Please contact Technical Service for linearity of dilution.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sooah Jang et al.
Psychiatry investigation, 15(2), 205-213 (2018-02-25)
Conventional methods for organotypic hippocampal tissue slice culture (OHSC) have shown several disadvantages or limitations regarding age of animals used, duration of culture and difficulty using neurodegenerative models. Therefore, we tried to establish OHSC from old 3xTg-Alzheimer's disease (AD) mice
Teresa Niccoli et al.
Current biology : CB, 26(17), 2291-2300 (2016-08-16)
Glucose hypometabolism is a prominent feature of the brains of patients with Alzheimer's disease (AD). Disease progression is associated with a reduction in glucose transporters in both neurons and endothelial cells of the blood-brain barrier. However, whether increasing glucose transport
Ekram Abdel Salam et al.
Acta myologica : myopathies and cardiomyopathies : official journal of the Mediterranean Society of Myology, 33(1), 13-18 (2014-05-21)
The presence of non-progressive cognitive impairment is recognized as a common feature in a substantial proportion of patients with Duchenne muscular dystrophy (DMD). Concurrently, the amyloid beta peptide (Aβ42) protein has been associated with changes in memory and cognitive functions.
Kendra L Puig et al.
Journal of Alzheimer's disease : JAD, 44(4), 1263-1278 (2014-11-20)
Alzheimer's disease (AD) is a neurodegenerative disorder histologically characterized by amyloid-β (Aβ) protein accumulation and activation of associated microglia. Although these features are well described in the central nervous system, the process and consequences of Aβ accumulation in the enteric
Ruth Maron et al.
International journal of molecular sciences, 21(9) (2020-05-10)
The two major proteins involved in Alzheimer's disease (AD) are the amyloid precursor protein (APP) and Tau. Here, we demonstrate that these two proteins can bind to each other. Four possible peptides APP1 (390-412), APP2 (713-730), Tau1 (19-34) and Tau2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service