Skip to Content
Merck
All Photos(1)

Documents

EZGLU

Millipore

Multi Species Glucagon ELISA Kit

detects and quantifies Glucagon in 20 μL serum or plasma

Synonym(s):

Glucagon assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

product name

Glucagon ELISA Kit, Chemiluminescent,

species reactivity

porcine, human, mouse, rat

Quality Level

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr assay)

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum

application(s)

research use

assay range

inter-assay cv: <10%
intra-assay cv: <5%
sensitivity: .003  ng/mL
standard curve range: 0.02 -2 ng/mL
accuracy: 101%

detection method

chemiluminescent (425nm)

shipped in

wet ice

storage temp.

2-8°C

General description

Glucagon is the most important counter-regulatory hormone to insulin in blood glucose homeostasis. It elevates blood glucose levels by stimulating gluconeogenesis and glycogenolysis while inhibiting glycogenesis and gylcolysis in the liver. This hormone is extremely important in studying therapeutic targets for diabetes, particularly Type I.

Specificity

Glucagon (Human, Mouse, Rat and Porcine) 100%
Oxyntomodulin (Human, Mouse and Rat) <5%
Glucagon (1-18) 0%
Glucagon 19-29 (mini Glucagon) 0%

Used to detect/quantify: Glucagon

Application

This assay is a chemiluminescent sandwich ELISA & is based on the capture of glucagon molecules in the sample, after extraction & reconstitution.

Research Category: Metabolism
Research Sub-Category: Obesity, Metabolic Disorders

Packaging

96-well strip plate

Other Notes

Contact Technical Service for this information

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service