EZFREAD01CN
EZ-Fluo™ Reader
Contains EZ-Fluo™ reader, power cord for China
Synonym(s):
Detection instrument, EZ-Fluo™ Reader Kit (CN), For rapid detection of microbial contamination in filterable samples, suitable for bioburden testing
About This Item
AC/DC input
100 V / 240 V, 50 - 60 Hz
Quality Level
manufacturer/tradename
EZ-Fluo™
parameter
4.4 kg at 15 - 40 °C
H × W × L
12.5 cm × 14.2 cm × 24.9 cm
application(s)
environmental
food and beverages
pharmaceutical
compatibility
EZ-Fluo™ Rapid Microbial Detection System
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service