Skip to Content
Merck
All Photos(5)

Key Documents

EZCURVE01

Millipore

EZ-PAK® Membrane Dispenser Curve

Membrane filtration, General microbiological analysis

Synonym(s):

EZ-CURVE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
Pricing and availability is not currently available.

Quality Level

manufacturer/tradename

EZ-Pak®

packaging

pkg of (1 unit)

dilution

(Membrane filter dispenser)

input

sample type beer
sample type bottled water
sample type pharmaceutical(s)
sample type cosmetics

application(s)

bioburden testing
water monitoring
(Beverages, Water)

compatibility

Membrane filtration
for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Related Categories

General description

EZ-PAK® Membrane Dispenser Curve is a robust, durable, and high-throughput membrane dispenser. It is lightweight, portable, and can be easily moved between lab benches. The dispenser provides high-speed, sterile dispensing with the no-touch operation. EZ-PAK® membranes are the consumables loaded on the dispenser for easy handling. For better efficiency and convenience, pair-up with our MICROFIL® filtration system is recommended. Each cartridge contains 150 membranes. The membranes can be unwrapped, dispensed, and consistently well-positioned for transfer with forceps, without any touch. Software controls prevent the accidental dispensing of multiple membranes. The smooth shape facilitates easy cleaning, and the plastic material is compatible with a large range of disinfection agents. The new design with cover allows for easy loading of cartridge in less than 30 seconds. The industrial infrared sensor enables the dispense of a membrane in less than one second. The dispenser can work with or without connection to a power outlet. Up to 10,000 membranes can be dispensed, with a fully charged Lithium-ion battery, and a standby mode is activated to save energy after 10 minutes of idle time.

Application

EZ-PAK® Membrane Dispenser Curve is used for efficient and consistent dispensing of membrane filters for bioburden testing.

Features and Benefits

  • High-speed sterile membrane dispensing
  • No-touch operation
  • One-handed membrane transfer

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

Related Content

A touch-free membrane filter dispenser can streamline your membrane filtration testing workflow. Use with EZ-Pak® filter membranes for high-volume sampling.

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service