This primary denaturant is methanol, however, other components are also added.
See the link below to review the SDS, section 3: https://www.sigmaaldrich.com/sds/mm/ex0278
EX0278
Ethanol (denatured)
Non UV, OmniSolv®
Synonym(s):
Alcohol, Ethanol, Denatured
About This Item
Recommended Products
Product Name
Ethyl Alcohol, Non UV, denatured OmniSolv®
Quality Level
form
liquid
impurities
≤0.5% Water
≤5 ng/L Capillary ECD responsive substances (as PCNB)
color
APHA: ≤10
transition temp
flash point 11 °C
shipped in
ambient
storage temp.
room temp
SMILES string
OCC
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Physical form
Preparation Note
Analysis Note
Color (APHA): 10 max
Filtered through 0.2 m filter: To pass test
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Water (H2O): 0.5% max
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
51.8 °F
Flash Point(C)
11 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is this denatured with? Thnaks
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service