Skip to Content
Merck
All Photos(1)

Key Documents

EST0000L06GP5

Millipore

Mobius® Essential Assembly Tubing

AseptiQuik® G Connector Manifold Tubing Assembly

Synonym(s):

Mobius® Essential Assembly, Tubing, AG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103801
Pricing and availability is not currently available.

material

C-Flex® tubing

Quality Level

Agency

complies with ISO 9001

product line

Mobius® Essential

technique(s)

buffer exchange: suitable
media fill: suitable

application(s)

cell therapy
gene therapy
life science and biopharma
liquid processing
mAb
ophthalmics
parenterals
pharma/biopharma processes
viral therapy
waste management

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Mobius® Essential Assemblies are standard ready-to-ship bag and tubing assemblies.

Application

Liquid Processing Applications

Features and Benefits

Standard products eliminate design and manufacturing lead times

Easily connects to other single-use assemblies

Ready-to-download Emprove® Dossier with comprehensive product and regulatory informatio

Packaging

Tubing Assemblies in 5 Pack

Components

C-Flex® 374 Tubing 1/2" ID x 3/4" OD, 6" long
AseptiQuik G Connector, 1/2" Hub

Legal Information

AseptiQuik is a registered trademark of Colder Products Company
C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Mobius® Essential Assemblies offer simple, ready-to-ship bioprocess solutions; integrated into workflow with various bag sizes and tubing designs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service