The EF5 dosage required for your samples can vary based on the specific experimental conditions. According to the data sheet provided, for in vivo assays, it is recommended to include a no EF5 injection control and a competed stain antibody control for small animal studies. The EF5 dosage involves making a tail-vein injection using a specific concentration of the drug relative to the animal's weight and freezing tissues in OCT until ready to section.
EF5-30A4M
EF5 Hypoxia Detection Kit, Alexa Fluor™ 488
Synonym(s):
Green fluorescent hypoxia detection
About This Item
Recommended Products
technique(s)
immunofluorescence: suitable
immunohistochemistry: suitable
Quality Level
detection method
fluorometric
General description
Advantages of the EF5 Hypoxia Detection Kit:
• EF5 exists in only one form:
Pimidozole, an alternative hypoxia marker, exists in two forms; one of which is charged and very hydrophilic, the other lipophilic. Pimidozole thus has a very complex biodistribution. In contrast, EF5 is lipophilic and uncharged and this allows very rapid and even tissue distribution.
• EF5 binding images can be calibrated to provide quantitative data on the pO2 values of each cell (Koch CJ, 2002):
The fluorescent images obtained from EF5 binding can be calibrated according to camera settings and a “cube-binding” value which is obtained through a separate procedure. The intensity values of calibrated images are directly related to actual tissue pO2 values. As a result, these images provide information regarding not only where hypoxic areas may or may not be, but also data regarding the distribution and levels of hypoxia.
Reference:
Koch CJ (2002) Measurement of absolute oxygen levels in cells and tissues using oxygen sensors and 2-nitroimidazole EF5. Methods in Enzymology 352: 3-31.
Application
Cancer
Components
Anti-EF5, clone ELK3-51 Alexa Fluor™ 488 conjugate, 250 ug: (Part No. CS222729). 1 vial containing 125 uL mouse monoclonal antibody, provided at 2 mg/mL and formulated in PBS, 0.1% sodium azide, 30% glycerol and 1.5% BSA. Store at -20°C.
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Hypoxia detection assays to measure oxygen levels in both live and fixed cells and tissues.
-
Hi, I'm wondering how many normal-sized mice the EF5-30A4 kit would work for?
1 answer-
Helpful?
-
-
What is the recommended waiting period after injecting the animal?
1 answer-
For IV injection in a mouse model, a waiting period of approximately 10 minutes should be adequate. However, for IP injection, it is recommended to wait for at least 30 minutes, as per literature information. This is supported by a study in Neuroscience (1999; 89(4):1357-66) where EF5 solution was administered intraperitoneally 30 minutes prior to each treatment.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service