Skip to Content
Merck
All Photos(1)

Documents

ECM509

Sigma-Aldrich

QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), fluorimetric

The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with fluorescent detection.

Synonym(s):

cell migration assay, fluorometric chemotaxis assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.32

Quality Level

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®
QCM

technique(s)

activity assay: suitable
cell based assay: suitable

detection method

fluorometric

shipped in

wet ice

Application

Research Category
Cell Structure
The CHEMICON® QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. Each Chemicon Cell Migration Assay Kit contains sufficient reagents for the evaluation of 24 samples. The quantitative nature of this assay is especially useful for screening of pharmacological agents.

The CHEMICON QCM 24-well Migration Assay is intended for research use only; not for diagnostic applications.
The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with fluorescent detection.

Packaging

24 wells

Components

Sterile 24-well Cell Migration Plate Assembly: (Part No. 90333) Two 24-well plates with 12 inserts per plate (24 inserts total/kit).

Cell Detachment Solution: (Part No. 90131) One bottle - 16 mL.

4X Cell Lysis Buffer: (Part No. 90130) One bottle - 16 mL.

CyQUANTâ GR Dye1: (Part No. 90132) One vial - 75 μL

Forceps: (Part No. 10203) One each.

Storage and Stability

Store kit materials at 2-8°C for up to their expiration date. Do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah K Abey et al.
BBA clinical, 7, 23-35 (2017-01-06)
Stress has demonstrated effects on inflammation though underlying cell-cell communication mechanisms remain unclear. We hypothesize that circulating RNAs and extracellular vesicles (EVs) in patients with chronic stress contain signals with functional roles in cell repair. Blood transcriptome from patients with
Zhisheng Long et al.
Cancer cell international, 20, 481-481 (2020-10-13)
Circular RNAs (circRNAs) are important regulators in the pathogenesis of diseases and affects the occurrence and development of diseases. However, the role of circRNAs in osteosarcoma (OS) has not been fully elucidated. The expression of circ_0000285, miR-409-3p and insulin-like growth
Xi Xi Lai et al.
Molecular medicine reports, 17(2), 2472-2478 (2017-12-06)
Breast cancer is one of the most common types of carcinoma in humans. The aim of the present study was to identify the role of Notch 1 in the proliferation and invasion of human breast cancer cells. Firstly, the levels
Yukuan Feng et al.
European journal of cancer (Oxford, England : 1990), 47(15), 2353-2363 (2011-06-18)
Vascular endothelial growth factor C (VEGF-C) expression is associated with the malignant tumour phenotype making it an attractive therapeutic target. We investigated the biological roles of VEGF-C in tumour growth, migration, invasion and explored the possibility of VEGF-C as a
Dang Chao et al.
Oncology reports, 45(6) (2021-04-14)
Stomatin‑like protein 2 (SLP‑2) is associated with poor prognosis in several types of cancer, including pancreatic cancer (PC); however, the molecular mechanism of its involvement remains elusive. The present study aimed to elucidate the role of this protein in the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service