Skip to Content
Merck
All Photos(2)

Documents

E5SU871

Millipore

NovaSeptum® GO Preloaded System with Bottle, 5-port

sterile; γ-irradiated

Synonym(s):

NovaSeptum® GO Bottle, Sterile_Sampling_Container

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

material

316 stainless steel cannula (ASTM®)
PETG bottle
TPE tubing
polyester body (platinum-cured)
polyethylene bottle
silicone septum (platinum-cured)

Quality Level

Agency

according to ISO 11137 (sterilization)
according to ISO 146441
meets USP 88 Biological Reactivity (all component materials)

sterility

sterile; γ-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

-80-50 °C temp. range (-112-122 °F)
0.50 bar max. pressure (7.25 psi)

impurities

<2.15 EU/device bacterial endotoxins (LAL test)

fitting

outlet tubing (cap)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Device configuration: single unit sampling - preloaded system
The NovaSeptum® GO bottle sampling unit is intended for applications where high purity, such as low endotoxin levels, is required. This sampling unit is available with a 2 mm needle and must be used with a NovaSeptum® GO holder. The bio-neutral plastic bottles are ideal for all tests and available in a range of sizes. A closed, sterile sampling method can significantly reduce risk of cross contamination. Ideal for sampling from aseptic and sterile processes, the NovaSeptum® GO sterile sampling system provides consistently representative samples, safely and securely.

Application

sterile sampling

Packaging

E5SU871-80060 (5x60 ml bottles, 6 units per box)
E5SU871-80125 (5x125 ml bottles, 4 units per box)
E5SU871-80250 (5x250 ml bottles, 3 units per box)
E5SU871-80500 (5x500 ml bottles, 2 units per box)

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (bottle): Polyethylene Terephtalate Glycol
  • Tubing: Thermoplastic elastomer (TPE)
  • Outlet Tubing: Cap

Preparation Note

Units are integrity tested at regular intervals during manufacturing.
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service