Skip to Content
Merck
All Photos(1)

Documents

DX0145

Sigma-Aldrich

Dextrose

anhydrous Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

D-(+)-Glucose, D-Glucose, Corn sugar, Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-075-1

form

solid

color

white

mp

150-152 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

GR ACS Dextrose, Anhydrous
DX0145-3 2.5kg Poly Bottle

Application

anhydrous Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Appearance (description)powderAppearance of solutionpasses testChloride (Cl)0.01% maxColorWhiteHeavy metals (as Pb)0.0005% maxInsoluble matter0.005% maxIron (Fe)0.0005% maxLoss on drying0.2% maxSpecific rotation [a]25/D (10%, water)+52.5-+53.0degreesStarchpasses testSulfate and sulfite (as SO4)0.005% maxSulfated ash0.02% maxTitratable free acid0.002meq/g max

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service