Skip to Content
Merck

Skip To

DPLUS-100

DirectLoad Plus 100bp DNA Ladder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
41106514
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid (Reagent)

Quality Segment

packaging

vial of 500 μL

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

Northern blotting: suitable, Southern blotting: suitable

shipped in

dry ice

storage temp.

−20°C

Features and Benefits

It′s suitable for use as an electrophoresis marker for DNA

Preparation Note

-20 °C for up to 24 months, or 4 °C up to 12 months or room temperature for up to six months.

Other Notes

DirectLoad Plus 100bp DNA Ladder contains 11 fragments, which are obtained from PCR products and double-stranded DNA digested with appropriate restriction enzymes equilibrated to 10 mM Tris-HCl (pH 8.0) and 10 mM EDTA after phenol extraction.
The DNA includes fragments ranging from 100-1,500 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points.

Legal Information

DirectLoad is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DPLUS-100HDPLUS-1KDPLUS-1KS
technique(s)

Northern blotting: suitable, Southern blotting: suitable

technique(s)

Northern blotting: suitable, Southern blotting: suitable

technique(s)

Northern blotting: suitable, Southern blotting: suitable

technique(s)

Northern blotting: suitable, Southern blotting: suitable

form

liquid (Reagent)

form

liquid (Reagent)

form

liquid (Reagent)

form

-

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

growth mode

N/A

growth mode

N/A

growth mode

N/A

growth mode

N/A


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield






Global Trade Item Number

SKUGTIN
DPLUS-10004065268021668

Questions

Reviews

No rating value

Active Filters