Skip to Content
Merck
All Photos(1)

Documents

CX1057

Supelco

Chloroform

new

anhydrous, septum-sealed bottle DriSolv

Synonym(s):

Trichloromethane, Methylidyne trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-663-8

vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

solubility

8.7 g/L

density

1.492 g/mL at 25 °C (lit.)

shipped in

ambient

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Stabilized with AlkeneDriSolv solvents are packed under Nitrogen and sealed with a septum.
anhydrous, septum-sealed bottle DriSolv

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99% minColor (APHA)10 maxEvaporation residue3ppm maxFiltered through 0.2 µm filterTo pass testIdentity (IR-spectrum)ConformsWater (H2O)50ppm max

Legal Information

DRISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service