CPMB02202
Ion-Ex Polishing Cartridge
L 55 cm (22 in.), for use with Super-Q® Ultrapure High Flow Water Systems
Synonym(s):
Ion-Ex Polishing Cartridge 55cm (22")
About This Item
Recommended Products
material
(Nuclear grade mixed bed ion exchange resin)
Quality Level
feature
Removal of Inorganics and particulate contaminants
packaging
pkg of 2 units
L
55 cm (22 in.)
capacity
175 g ion exchange capacity (NaCl)
compatibility
for use with Super-Q® Ultrapure High Flow Water Systems
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Efficiently removes inorganic particulates and contaminants within the feed stream of the Super-Q® Plus lab water purification systems.
- Contains virgin ion-exchange resins.
- Consistently produces ultrapure water quality which exceeds Type I ASTM/CAP/CLSI specifications.
Legal Information
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service