CNHALA210
SteriSecure™ Device
For Ampoules and Bags, 10/pack
About This Item
material
PVC tubing
mixed cellulose esters (HA) membrane
stainless steel needle
styrene-acrylonitrile (SAN) (canister)
Quality Level
Agency
ChP
sterility
sterile; γ-irradiated
manufacturer/tradename
SteriSecure™
parameter
max. pressure at 3.15 bar and 25 °C
45 °C max. temp.
L
850 mm
pore size
0.45 μm (Canister membrane)
input
liquid
application(s)
pharmaceutical
sterility testing
compatibility
for use with Steritest® systems
General description
The closed system minimizes false positives and offers the highest level of quality and reliability. This device ensures that pharmaceutical products are never exposed to the environment during the testing process. This membrane provides an optimal filtration flow rate for standard products. This test system offers an optimized and fully regulatory compliant testing process when used with the Steritest® Symbio pump, specific accessories, and high-quality culture media and rinsing fluids.
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service