Skip to Content
Merck
All Photos(1)

Key Documents

CL07

Sigma-Aldrich

TMB Solution

This TMB Solution is validated for use in ELISA for the detection of TMB Solution.

Synonym(s):

TMB Solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41105333
NACRES:
NA.41
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze
protect from light

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Nc1c(cc(cc1C)c2cc(c(c(c2)C)N)C)C

InChI

1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3

InChI key

UAIUNKRWKOVEES-UHFFFAOYSA-N

General description

A soluble, non-toxic, non-flammable horseradish peroxidase substrate.

Application




ELISA

Warning

Toxicity: Harmful (C)

Physical form

In a mildly acidic buffer with proprietary stabilizers, pH 3.55 ± 0.20.

Other Notes

Product does not contain hazardous ingredients such as polar aprotic solvents (i.e., N,N-Dimethylformamide (DMF) or DMSO) that are found in other TMB formulations. There is also no alcohol in TMB (Soluble Substrate) so it is not flammable and does not require special handling for shipping. Unreacted TMB (Soluble Substrate) should be colorless or a very light-bluish-green prior to use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is there a usage protocol available for this product? How much hydrogen peroxide should be added to this reagent for use in ELISA?

    1 answer
    1. This product is a ready to use product. The addition of hydrogen peroxide is not required. For use in Elisa, please see the general protocol below:
      1. Complete all required incubations with antibodies and HRP-labeled probes.
      2. Wash plate wells at least 4 times with phosphate-buffered saline (PBS) or Tris-buffered saline (TBS), containing 0.1% Tween®-20 detergent.
      3. After the final wash, shake and blot all residual buffer from the wells.
      4. Add 100 µl TMB solution to each well and incubate for 5-30 min.
      5. Allow the substrate reaction to develop a blue soluble reaction by incubating for the optimal time determined for the assay.
      6. The reaction should be stopped by using a 450 nm or 650 nm stop solution.
      7. Measure the absorbance at 450 nm or 650 nm of each microwell within 1 hour.
      8. Options for measurements:
      a. For kinetic assays, the reaction can be monitored as a function of time by reading absorbance at 650 nm at intermediate intervals.
      b. For endpoint assays that preserve the blue chromogen, the reaction should be stopped by addition of 100 µl of 0.1% sodium fluoride (NaF) and the absorbance read at 650 nm.
      c. If increased sensitivity is desired for endpoint assays, the reaction should be stopped by addition of 100 µl of either 500 mM H2SO4 or 250 mM HCl and the absorbance read at 450 nm WITHIN 5 MIN. Addition of acid converts the blue radical to the yellow diimine, which absorbs at 450 nm.

      NOTE: Optimal incubation times may vary depending on the amount of HRP present. If color develops too quickly, zero-order kinetics will not prevail. Dilution of the probe, antibody, or HRP-labeled reagent may be required. Variations in time, reagent volumes, and temperature may also require further standardization by the user.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service