Skip to Content
Merck
All Photos(1)

Documents

CDRCKT060

Milli-Q

Reverse Osmosis Cartridge

Produces Reverse Osmosis water at a flow rate of 60 L/h, For use with Milli-Q® CLX / HR / HX systems

Synonym(s):

Reverse Osmosis Cartridge – 60 L/h

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

packaging

pkg of 1 unit

parameter

60 L/hr flow rate

compatibility

for use with Milli-Q® CLX Water Purification System
for use with Milli-Q® HR Water Purification System
for use with Milli-Q® HX SD Water Purification System

Looking for similar products? Visit Product Comparison Guide

General description

The reverse osmosis (RO) cartridge removes 95%-99% of contaminants including ions, particulates, bacteria and organic molecules from potable feed water.

Application

RO water is used in laboratories for applications such as general rinsing, feedwater to washing machines and autoclaves, basic reagent preparation, or as a feed to ultrapure water purification systems.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service