Skip to Content
Merck
All Photos(1)

Key Documents

CBL490

Sigma-Aldrich

Anti-Integrin αV (ITGAV/CD51) Antibody

CHEMICON®, mouse monoclonal, 13C2

Synonym(s):

CD51

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Integrin αV Antibody, clone 13C2, clone 13C2, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13C2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Related Categories

Application

Anti-Integrin αV Antibody, clone 13C2 is an antibody against Integrin αV for use in FC, IH.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lalit Kumar Sharma et al.
Nature communications, 9(1), 4399-4399 (2018-10-26)
Pantothenate kinase (PANK) is a metabolic enzyme that regulates cellular coenzyme A (CoA) levels. There are three human PANK genes, and inactivating mutations in PANK2 lead to pantothenate kinase associated neurodegeneration (PKAN). Here we performed a library screen followed by
Richard H Kimura et al.
PloS one, 6(2), e16112-e16112 (2011-03-03)
The Ecballium elaterium trypsin inhibitor (EETI-II), a 28-amino acid member of the knottin family of peptides, contains three interwoven disulfide bonds that form multiple solvent-exposed loops. Previously, the trypsin binding loop of EETI-II has been engineered to confer binding to
Jakob Dalgaard et al.
Journal of leukocyte biology, 77(5), 689-698 (2005-02-25)
CD11c+ dendritic cells (DC) and plasmacytoid DC (PDC) are the two major DC subsets in human peripheral blood. For the purpose of immunotherapy with DC, it is important to investigate the phagocytosis of killed tumor cells by different DC subsets.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service