Skip to Content
Merck
All Photos(1)

Key Documents

CBL458F

Sigma-Aldrich

Anti-CD49f Antibody, FITC conjugated, clone 4F10, FITC conjugated

clone 4F10, Chemicon®, from mouse

Synonym(s):

Integrin alpha6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA6(3655)

Related Categories

Specificity

This antibody is specific to CD49f (integrin alpha6) chain) which is expressed on epithelial and endothelial basement membrane. The VLA alpha6 chain, which consists of a single disulfide linked 120 kDa subunit and a small doublet subunit of 25/30 kDa, is noncovalently-associated with the VLA beta-chain (CD29) in VLA-6 or alpha6/beta1 complex or with integrin beta4 chain (alpha6/beta4). The antigen is expressed on platelets, monocytes, thymocytes and T lymphocytes.

FUSION PARTNER: NS1 myeloma cell line

Immunogen

Cell line SW/222 (Human colorectal carcinoma).

Application

Anti-CD49f Antibody, FITC conjugated, clone 4F10, FITC conjugated is an antibody against CD49fd for use in FC, IP, IH.
Immunohistological studies on frozen sections

Immunoprecipitation of the laminin receptor

Flow Cytometry: 10 μL per 106

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Linkage

Replaces: MAB2254F

Physical form

Protein A purified immunoglobulin conjugated to fluorescein isothiocyanate (FITC). Liquid in PBS containing 1mg/mL BSA and 10mM sodium azide.

Storage and Stability

Maintain at 2-8°C for up to 12 months from date of receipt, protected from light. DO NOT FREEZE.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service