Skip to Content
Merck
All Photos(1)

Documents

BX1791

Supelco

Isobutyl Alcohol

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Isobutyl Alcohol, Isobutanol, 2-Methyl-1-propanol, 2-methyl-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
MDL number:
UNSPSC Code:
12191502
EC Index Number:
201-148-0

Agency

meets USP/NF

Quality Level

vapor pressure

12 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

impurities

≤0.0005 meq/g Titrable acid
≤0.01% Carbonyl compounds (as butyraldehyde)
≤0.02% Carbonyl compounds (as 2-butanone)
≤0.1% Water (H2O)

evapn. residue

≤0.001%

color

APHA: ≤10

pH

7 (20 °C, 80 g/L in H2O)

bp

106-108 °C/1013 hPa

mp

-108 °C

transition temp

flash point 28 °C

solubility

85 g/L

density

0.802 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (GC): 99.0% min
Carbonyl compounds (as 2-butanone): 0.02% max
Carbonyl compounds (as butyraldehyde): 0.01% max
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Solubility in water: To pass test
Titrable acid: 0.0005meq/g max
Water (H2O): 0.1% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service