Skip to Content
Merck
All Photos(1)

Key Documents

BX1735

Supelco

Butyl acetate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
MDL number:
UNSPSC Code:
12191502
EC Index Number:
204-658-1
grade:
reagent
bp:
124-126 °C (lit.)
vapor pressure:
15 mmHg ( 25 °C)
8 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

Agency

meets USP/NF

vapor density

4 (vs air)

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

790 °F

expl. lim.

7.6 %

dilution

(for analytical testing)

impurities

≤0.0016 meq/g Titrable acid
≤0.1% Water (H2O)
≤0.2% n-Butyl alcohol

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.394 (lit.)

pH

6.2 (20 °C, 5.3 g/L in H2O, External MSDS)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

transition temp

flash point 27 °C

solubility

5.3 g/L

density

0.88 g/cm3 at 20 °C
0.88 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BX1735-6 is packaged in poly coated glass bottles
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Assay (GC): 99.5% min
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Substances darkened by sulfuric acid: To pass test
Titrable acid: 0.0016meq/g max
Water (H2O): 0.1% max
n-Butyl alcohol: 0.2% max

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service