Skip to Content
Merck
All Photos(1)

Key Documents

AR002

Sigma-Aldrich

Freund′s Incomplete Adjuvant

Synonym(s):

Freund′s Adjuvant, Incomplete Freund′s Adjuvant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

form

liquid

Quality Level

manufacturer/tradename

Chemicon®

shipped in

wet ice

Application

Composition:

85% Drakeol 6VR (Mineral Oil)

15% Mannide Monooleate (Arlacel A)
Research Category
All

Storage and Stability

Store at 2-8°C for up to twelve months.

Note: This material is of U.S. origin and donor animals were free from visible disease at the time of blood collection.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E U Sumer et al.
Osteoarthritis and cartilage, 15(2), 212-221 (2006-09-26)
Aggrecan is the major proteoglycan in articular cartilage and is known to be degraded by various proteases, including matrix metalloproteinases (MMPs). The present study was undertaken to develop immunoassays detecting aggrecan and its fragments generated by MMP and non-MMP-mediated proteolysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service