For use in blocking the staining of Chemicon′s catalog numbers AB143, AB144P and AB144. Suggested concentration 2-5 μg/mL. AG220 should be incubated with the antibody for 3 hours at +2-8°C prior to being used on the tissue sections.
Enzyme is denatured by heat, but its activity level has not been determined. Residual activity may be present.
Optimal working dilution must be determined by the end user.
Research Category Neuroscience
Research Sub Category Neurotransmitters & Receptors
Neuronal & Glial Markers
Physical form
Liquid in 50 mM KPO4 buffer pH 7.5 containing 10% glycerol and 0.05M NaCl. Contains no preservative.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of comparative neurology, 519(12), 2335-2352 (2011-04-02)
Neurotrophins acting through high-affinity tyrosine kinase receptors (trkA, trkB, and trkC) play a crucial role in regulating survival and maintenance of specific neuronal functions after injury. Adult motoneurons supplying extraocular muscles survive after disconnection from the target, but suffer dramatic
Nuclear organization of cholinergic, putative catecholaminergic and serotonergic nuclei in the brain of the eastern rock elephant shrew, Elephantulus myurus.
Raymond P Pieters,Nadine Gravett,Kjell Fuxe,Paul R Manger
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.