Skip to Content
Merck
All Photos(2)

Key Documents

ABT335

Sigma-Aldrich

Anti-SYNPO2L Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Synaptopodin 2-like, cytoskeletal heart-enriched actin-associated protein, CHAP, SYNPO2L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYNPO2L(79933)

General description

SYNPO2L, also known as Synaptopodin 2-like protein, is a member of the synaptopodin family. SYNPO2L was initially identified as a novel heart-enriched gene that encodes a cytoskeletal protein highly expressed in the Z-disc of heart and skeletal muscle. It has been shown that even though SYNPO2L is primarily localized to the sarcomere, it can also translocate to the nucleus. Recent studies in zebrafish suggest that SYNPO2L may play an important role in muscle development.

Immunogen

KLH-conjugated linear peptide corresponding to human SYNPO2L.

Application

Anti-SYNPO2L Antibody is a highly specific rabbit polyclonal antibody, that targets Synaptopodin 2-like protein & has been tested in western blotting, Immunofluorescence & IHC.
Immunofluorescence Analysis: 5 µg/mL from a representative lot detected SYNPO2L in mouse skeletal muscle cells.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected SYNPO2L in human cardiac myocytes.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blotting in human thymus tissue lysate.

Western Blotting Analysis: 1µg/mL of this antibody detected SYNPO2L in 15 µg of human thymus tissue lysate.

Target description

~95 kDa observed. Uncharacterized bands may appear in some lysates. Uniprot describes 2 isoforms at ~79 kDa and ~103 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human thymus tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service